4-Bromo-7-(trifluoromethyl)-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

19881-1G 19881-25G 19881-500MG 19881-5G
76926-902EA 182.48 USD
76926-902 76926-904 76926-906 76926-908
4-Bromo-7-(trifluoromethyl)-1H-indazole ≥97%
4-Bromo-7-(trifluoromethyl)-1H-indazole
Formula: C₈H₄BrF₃N₂
MW: 265.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16658657
CAS Number: 1186334-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR