4-Fluoro-3-(Methylsulfonyl)Phenylboronic Acid Pinacol Ester ≥97%

Supplier: AOBChem USA

Warning

19851-1G 19851-25G 19851-500MG 19851-5G
76926-884EA 290.69 USD
76926-884 76926-886 76926-888 76926-890
4-Fluoro-3-(Methylsulfonyl)Phenylboronic Acid Pinacol Ester ≥97%
4-Fluoro-3-(Methylsulfonyl)Phenylboronic Acid Pinacol Ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BFO₄S
Storage Temperature: Ambient
MDL Number: MFCD18383936
CAS Number: 1203655-90-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR