1-(5-Bromo-2,4-difluorophenyl)ethanone ≥95%
Supplier: AOBChem USA
|
Warning
|
19793-25G
19793-5G
76926-864EA
310.85
USD
76926-864
76926-866
1-(5-Bromo-2,4-difluorophenyl)ethanone ≥95%
1-(5-Bromo-2,4-difluorophenyl)ethanone
Formula:
C₈H₅BrF₂O Storage Temperature: Ambient |
MDL Number:
MFCD13190613 CAS Number: 864773-64-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...