4-Bromo-3-(methoxycarbonyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

19738-1G 19738-25G 19738-500MG 19738-5G
76926-854EA 186.72 USD
76926-854 76926-856 76926-858 76926-860
4-Bromo-3-(methoxycarbonyl)phenylboronic acid ≥97%
4-Bromo-3-(methoxycarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BBrO₄
Storage Temperature: Refrigerator
MDL Number: MFCD23379501
CAS Number: 870196-03-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR