5-Chloro-2-iodo-m-xylene ≥97% research grade

Supplier: AOBChem USA

Warning

19723-1G 19723-25G 19723-500MG 19723-5G
76926-840EA 242.95 USD
76926-840 76926-842 76926-844 76926-846
5-Chloro-2-iodo-m-xylene ≥97% research grade
5-Chloro-2-iodo-m-xylene
Formula: C₈H₈ClI
Storage Temperature: Refrigerator
MDL Number: MFCD11846954
CAS Number: 404337-42-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR