4-Chloro-3,5-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

19692-1G 19692-25G 19692-500MG 19692-5G
76926-826EA 124.13 USD
76926-826 76926-828 76926-830 76926-832
4-Chloro-3,5-dimethylbenzoic acid ≥97%
4-Chloro-3,5-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
Storage Temperature: Refrigerator
MDL Number: MFCD11111063
CAS Number: 90649-78-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR