(5-Bromo-2-chlorophenyl)boronic acid ≥95%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
5-Bromo-2-chlorophenylboronic acid
19552-1G
19552-25G
19552-500MG
19552-5G
76926-790EA
439.22
USD
76926-790
76926-792
76926-794
76926-796
(5-Bromo-2-chlorophenyl)boronic acid ≥95%
(5-Bromo-2-chlorophenyl)boronic acid
Formula:
C₆H₅BBrClO₂ MW: 235.27 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD12026740 CAS Number: 774608-50-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...