4-Bromo-3-fluorothioanisole ≥95%

Supplier: AOBChem USA

Warning

19365-1G 19365-25G 19365-5G
76926-756EA 96.55 USD
76926-756 76926-758 76926-760
4-Bromo-3-fluorothioanisole ≥95%
4-Bromo-3-fluorothioanisole

Precaución: for Research Use Only

Formula: C₇H₆BrFS
Storage Temperature: Refrigerator
MDL Number: MFCD09027283
CAS Number: 917562-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR