1-Benzyl-5-bromo-1H-indazole ≥97% research grade

Supplier: AOBChem USA

Warning

19293-25G 19293-5G
76926-734EA 519.85 USD
76926-734 76926-736
1-Benzyl-5-bromo-1H-indazole ≥97% research grade
1-Benzyl-5-bromo-1H-indazole
Formula: C₁₄H₁₁BrN₂
Storage Temperature: Ambient
MDL Number: MFCD20526373
CAS Number: 1087160-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR