4-Chlor-3-(isopropylcarbamoyl)phenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 4-Chloro-3-(isopropylcarbamoyl)benzeneboronic acid, 4-Chloro-3-(isopropylcarbamoyl)phenylboronic acid, (4-Chloro-3-(isopropylcarbamoyl)phenyl)boronic acid

19240-1G 19240-25G 19240-5G
76926-712EA 76.39 USD
76926-712 76926-714 76926-716
4-Chlor-3-(isopropylcarbamoyl)phenylboronic acid ≥95%
4-Chlor-3-(isopropylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BClNO₃
MW: 241.48 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07783866
CAS Number: 871332-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR