5-Bromo-2-methoxy-3-pyridineboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-2-methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 5-Bromo-2-methoxypyridine-3-boronic acid, pinacol ester

19182-25G 19182-5G
76926-696EA 460.44 USD
76926-696 76926-698
5-Bromo-2-methoxy-3-pyridineboronic acid pinacol ester ≥97%
5-Bromo-2-methoxy-3-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BBrNO₃
MW: 313.99 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07781157
CAS Number: 1073353-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR