5-Amino-2-fluorobenzeneboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-FLUORO-3-(4,4,5,5-TETRAMETHYL-1,3,2-DIOXABOROLAN-2-YL)ANILINE, 5-AMINO-2-FLUOROPHENYLBORONIC ACID PINACOL ESTER, 5-Amino-2-fluorophenylboronic acid pinacol ester

19153-1G 19153-25G 19153-500MG 19153-5G
76926-686EA 161.26 USD
76926-686 76926-688 76926-690 76926-692
5-Amino-2-fluorobenzeneboronic acid pinacol ester ≥97%
5-Amino-2-fluorobenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BFNO₂
Storage Temperature: Ambient
MDL Number: MFCD16996293
CAS Number: 1152441-29-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR