5-Fluoro-2-iodophenol ≥95% research grade

Supplier: AOBChem USA

Warning

Synonyms: 5-Fluoro-2-iodo-phenol

19035-1G 19035-25G 19035-500MG 19035-5G
76926-660EA 147.47 USD
76926-660 76926-662 76926-664 76926-666
5-Fluoro-2-iodophenol ≥95% research grade
5-Fluoro-2-iodophenol
Formula: C₆H₄FIO
Storage Temperature: Refrigerator
MDL Number: MFCD16999912
CAS Number: 186589-87-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR