5-Bromo-2-(cyclopropylmethoxy)pyridine ≥97%

Supplier: AOBChem USA

Warning

18968-1G 18968-25G 18968-5G
76926-644EA 110.34 USD
76926-644 76926-646 76926-648
5-Bromo-2-(cyclopropylmethoxy)pyridine ≥97%
5-Bromo-2-(cyclopropylmethoxy)pyridine
Formula: C₉H₁₀BrNO
MW: 228.08 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13190758
CAS Number: 494772-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR