5-Bromo-2-ethoxy-3-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

18876-1G 18876-25G 18876-500MG 18876-5G
76926-610EA 177.18 USD
76926-610 76926-612 76926-614 76926-616
5-Bromo-2-ethoxy-3-pyridineboronic acid ≥97%
5-Bromo-2-ethoxy-3-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₇H₉BBrNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD07363847
CAS Number: 871332-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR