5-Chloro-3-fluoropyridine-2-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

18795-1G 18795-25G 18795-500MG 18795-5G
76926-564EA 130.5 USD
76926-564 76926-566 76926-568 76926-570
5-Chloro-3-fluoropyridine-2-carboxylic acid ≥97%
5-Chloro-3-fluoropyridine-2-carboxylic acid
Formula: C₆H₃ClFNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD12827555
CAS Number: 207994-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR