5-Bromo-2,4-dichlorotoluene ≥97%

Supplier: AOBChem USA

Warning

18694-1G 18694-25G 18694-5G
76926-544EA 67.9 USD
76926-544 76926-546 76926-548
5-Bromo-2,4-dichlorotoluene ≥97%
5-Bromo-2,4-dichlorotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrCl₂
MW: 239.925 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09800823
CAS Number: 85072-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR