5-Chloro-2-methoxy-4-methylpyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
18613-1G
18613-25G
18613-5G
76926-518EA
62.6
USD
76926-518
76926-520
76926-522
5-Chloro-2-methoxy-4-methylpyridine ≥97%
5-Chloro-2-methoxy-4-methylpyridine
Precaución: for Research Use Only
Formula:
C₇H₈ClNO MW: 157.598 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD07375094 CAS Number: 851607-29-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...