4-Chloro-3,5-difluorophenylboronic acid, pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(4-Chloro-3,5-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

18581-1G 18581-25G 18581-500MG 18581-5G
76926-502EA 209 USD
76926-502 76926-504 76926-506 76926-508
4-Chloro-3,5-difluorophenylboronic acid, pinacol ester ≥97%
4-Chloro-3,5-difluorophenylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₄BClF₂O₂
MW: 274.499 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22418269
CAS Number: 1165935-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR