5-Bromo-2-ethoxypyridine-4-carboxaldehyde ≥97%

Supplier: AOBChem USA

Warning

18452-25G 18452-5G
76926-482EA 590.93 USD
76926-482 76926-484
5-Bromo-2-ethoxypyridine-4-carboxaldehyde ≥97%
5-Bromo-2-ethoxypyridine-4-carboxaldehyde

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
MW: 230.059 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16295251
CAS Number: 1224604-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR