5-(Benzyloxy)-2-bromopyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Benzyloxy-2-bromopyridine

18351-25G 18351-5G
76926-452EA 488.02 USD
76926-452 76926-454
5-(Benzyloxy)-2-bromopyridine ≥97%
5-(Benzyloxy)-2-bromopyridine
Formula: C₁₂H₁₀BrNO
MW: 264.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11874623
CAS Number: 630120-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR