5-Bromo-2-ethoxybenzotrifluoride ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-4-ethoxy-3-(trifluoromethyl)benzene, 4-Bromo-1-ethoxy-2-(trifluoromethyl)benzene

18343-25G 18343-5G
76926-446EA 215.37 USD
76926-446 76926-448
5-Bromo-2-ethoxybenzotrifluoride ≥97% research grade
5-Bromo-2-ethoxybenzotrifluoride
Formula: C₉H₈BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD08059521
CAS Number: 914635-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR