5-(Methoxycarbonyl)pyridine-3-boronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 5-borononicotinate, Methyl 5-boronopyridine-3-carboxylate, (5-(Methoxycarbonyl)pyridin-3-yl)boronic acid

18301-1G 18301-25G 18301-500MG 18301-5G
76926-422EA 124.13 USD
76926-422 76926-424 76926-426 76926-428
5-(Methoxycarbonyl)pyridine-3-boronic acid ≥95%
5-(Methoxycarbonyl)pyridine-3-boronic acid

Precaución: for Research Use Only

Formula: C₇H₈BNO₄
MW: 180.954 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07363742
CAS Number: 871329-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR