5-Dihydroxyboronylnicotinic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Carboxypyridine-3-boronic acid

18299-1G 18299-25G 18299-5G
76926-408EA 108.22 USD
76926-408 76926-410 76926-412
5-Dihydroxyboronylnicotinic acid ≥97%
5-Dihydroxyboronylnicotinic acid
Formula: C₆H₆BNO₄
MW: 166.93 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02323212
CAS Number: 913836-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR