(5-Formyl-2-methylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

18163-1G 18163-25G 18163-500MG 18163-5G
76926-386EA 203.7 USD
76926-386 76926-388 76926-390 76926-392
(5-Formyl-2-methylphenyl)boronic acid ≥97%
(5-Formyl-2-methylphenyl)boronic acid
Formula: C₈H₉BO₃
Storage Temperature: Ambient
MDL Number: MFCD16295249
CAS Number: 1186398-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR