4-Methoxy-2-(trifluoromethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

18014-25G 18014-5G
76926-352EA 524.09 USD
76926-352 76926-354
4-Methoxy-2-(trifluoromethoxy)benzaldehyde ≥97%
4-Methoxy-2-(trifluoromethoxy)benzaldehyde
Formula: C₉H₇F₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD06660317
CAS Number: 886503-52-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR