5-Benzyloxypyridine-3-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

17828-1G 17828-25G 17828-500MG 17828-5G
76926-302EA 141.1 USD
76926-302 76926-304 76926-306 76926-308
5-Benzyloxypyridine-3-carbaldehyde ≥97%
5-Benzyloxypyridine-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO₂
MW: 213.232 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18250511
CAS Number: 263270-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR