5-Bromo-2-isopropoxybenzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

17739-1G 17739-25G 17739-5G
76926-286EA 79.57 USD
76926-286 76926-288 76926-290
5-Bromo-2-isopropoxybenzaldehyde ≥97% research grade
5-Bromo-2-isopropoxybenzaldehyde
Formula: C₁₀H₁₁BrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD02257437
CAS Number: 138505-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR