4-Bromo-3-fluoro-N-methoxy-N-methyl-benzamide ≥95%

Supplier: AOBChem USA

Warning

17697-1G 17697-25G 17697-5G
76926-268EA 78.51 USD
76926-268 76926-270 76926-272
4-Bromo-3-fluoro-N-methoxy-N-methyl-benzamide ≥95%
4-Bromo-3-fluoro-N-methoxy-N-methyl-benzamide

Precaución: for Research Use Only

Formula: C₉H₉BrFNO₂
MW: 262.076 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08443760
CAS Number: 343564-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR