4-Fluoro-3-(phenylcarbamoyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

17443-1G 17443-25G 17443-5G
76926-194EA 62.6 USD
76926-194 76926-196 76926-198
4-Fluoro-3-(phenylcarbamoyl)phenylboronic acid ≥97%
4-Fluoro-3-(phenylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁BFNO₃
Storage Temperature: Ambient
MDL Number: MFCD08235052
CAS Number: 874219-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR