4-Bromo-N,N,2-trimethylaniline ≥97%

Supplier: AOBChem USA

Warning

17397-1G 17397-25G 17397-5G
76926-168EA 96.55 USD
76926-168 76926-170 76926-172
4-Bromo-N,N,2-trimethylaniline ≥97%
4-Bromo-N,N,2-trimethylaniline

Precaución: for Research Use Only

Formula: C₉H₁₂BrN
MW: 214.102 g/mol
Storage Temperature: Ambient
CAS Number: 50638-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR