4-Fluoro-(2-methoxycarbonyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Fluoro-2-methoxycarbonylphenylboronic acid, (4-Fluoro-2-(methoxycarbonyl)phenyl)boronic acid

17305-1G 17305-25G 17305-5G
76926-148EA 117.76 USD
76926-148 76926-150 76926-152
4-Fluoro-(2-methoxycarbonyl)phenylboronic acid ≥97%
4-Fluoro-(2-methoxycarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BFO₄
MW: 197.9561 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07363762
CAS Number: 871329-81-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR