4-Isopropoxy-2-methylphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

17273-1G 17273-25G 17273-5G
76926-134EA 62.6 USD
76926-134 76926-136 76926-138
4-Isopropoxy-2-methylphenylboronic acid ≥95%
4-Isopropoxy-2-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₅BO₃
Storage Temperature: Ambient
MDL Number: MFCD07363788
CAS Number: 871126-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR