5-Bromo-2-chloro-3-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

17264-25G 17264-5G
76926-126EA 279.02 USD
76926-126 76926-128
5-Bromo-2-chloro-3-methoxypyridine ≥97%
5-Bromo-2-chloro-3-methoxypyridine
Formula: C₆H₅BrClNO
Storage Temperature: Ambient
MDL Number: MFCD08234976
CAS Number: 286947-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR