5-Acetyl-2-methoxybenzaldehyd ≥97%
Supplier: AOBChem USA
|
Warning
|
17244-1G
17244-25G
17244-500MG
17244-5G
76926-112EA
374.5
USD
76926-112
76926-114
76926-116
76926-118
5-Acetyl-2-methoxybenzaldehyd ≥97%
5-Acetyl-2-methoxybenzaldehyd
Precaución: for Research Use Only
Formula:
C₁₀H₁₀O₃ MW: 178.185 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD19301502 CAS Number: 531-99-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...