1-Bromo-4-chloro-2-fluoro-5-methylbenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
17224-1G
17224-25G
17224-5G
76926-088EA
62.6
USD
76926-088
76926-090
76926-092
1-Bromo-4-chloro-2-fluoro-5-methylbenzene ≥97%
1-Bromo-4-chloro-2-fluoro-5-methylbenzene
Formula:
C₇H₅BrClF Storage Temperature: Refrigerator |
MDL Number:
MFCD09800685 CAS Number: 200190-87-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...