5-Fluoro-4-methoxy-2-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

17094-1G 17094-500MG 17094-5G
76926-058EA 501.81 USD
76926-058 76926-060 76926-062
5-Fluoro-4-methoxy-2-methylphenylboronic acid ≥97%
5-Fluoro-4-methoxy-2-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BFO₃
MW: 183.973 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22414589
CAS Number: 313667-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR