5-(tert-Butyldimethylsilyloxy)-2-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-[(tert-Butyldimethylsilanyl)oxy]-2-fluorobenzaldehyde

17046-1G 17046-25G 17046-500MG 17046-5G
76926-014EA 124.13 USD
76926-014 76926-016 76926-018 76926-020
5-(tert-Butyldimethylsilyloxy)-2-fluorobenzaldehyde ≥97%
5-(tert-Butyldimethylsilyloxy)-2-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₉FO₂Si
MW: 254.373 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17676576
CAS Number: 113984-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR