6-Fluoro-2,3-dimethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

17038-1G 17038-25G 17038-500MG 17038-5G
76926-002EA 126.25 USD
76926-002 76926-004 76926-006 76926-008
6-Fluoro-2,3-dimethoxybenzoic acid ≥97%
6-Fluoro-2,3-dimethoxybenzoic acid
Formula: C₉H₉FO₄
Storage Temperature: Refrigerator
MDL Number: MFCD13193706
CAS Number: 265670-72-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR