5-Bromo-2-fluoro-3-methoxycarbonylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

16982-1G 16982-25G 16982-500MG 16982-5G
76925-986EA 290.69 USD
76925-986 76925-988 76925-990 76925-992
5-Bromo-2-fluoro-3-methoxycarbonylphenylboronic acid ≥97%
5-Bromo-2-fluoro-3-methoxycarbonylphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₇BBrFO₄
Storage Temperature: Ambient
MDL Number: MFCD27578341
CAS Number: 2377612-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR