5-Bromo-2-nitrilephenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

16876-1G 16876-25G 16876-5G
76925-948EA 87 USD
76925-948 76925-950 76925-952
5-Bromo-2-nitrilephenylboronic acid pinacol ester ≥97%
5-Bromo-2-nitrilephenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₅BBrNO₂
MW: 307.979 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20923870
CAS Number: 863868-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR