4-Carboxy-2-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

16866-1G 16866-25G 16866-500MG 16866-5G
76925-932EA 132.62 USD
76925-932 76925-934 76925-936 76925-938
4-Carboxy-2-methoxyphenylboronic acid ≥97%
4-Carboxy-2-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₅
Storage Temperature: Refrigerator
MDL Number: MFCD10696665
CAS Number: 741699-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR