4-Bromo-2-fluoro-6-(trifluoromethyl)phenol ≥97% research grade

Supplier: AOBChem USA

Warning

16791-1G 16791-25G 16791-5G
76925-912EA 99.73 USD
76925-912 76925-914 76925-916
4-Bromo-2-fluoro-6-(trifluoromethyl)phenol ≥97% research grade
4-Bromo-2-fluoro-6-(trifluoromethyl)phenol
Formula: C₇H₃BrF₄O
MW: 258.99 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16295240
CAS Number: 1224604-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR