4-Carboxy-2-fluoroacetophenone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Acetyl-3-fluorobenzoic acid

16740-1G 16740-25G 16740-500MG 16740-5G
76925-896EA 277.96 USD
76925-896 76925-898 76925-900 76925-902
4-Carboxy-2-fluoroacetophenone ≥97%
4-Carboxy-2-fluoroacetophenone

Precaución: for Research Use Only

Formula: C₉H₇FO₃
MW: 182.148 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24039156
CAS Number: 1427082-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR