(4-Chloro-2-fluoro-3-methoxyphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Chloro-2-fluoro-3-methoxyphenylboronic acid

16678-1G 16678-25G 16678-5G
76925-864EA 62.6 USD
76925-864 76925-866 76925-868
(4-Chloro-2-fluoro-3-methoxyphenyl)boronic acid ≥97%
(4-Chloro-2-fluoro-3-methoxyphenyl)boronic acid
Formula: C₇H₇BClFO₃
MW: 204.39 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17015742
CAS Number: 944129-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR