5-Ethyl-2-fluorophenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

16549-1G 16549-25G 16549-5G
76925-828EA 94.43 USD
76925-828 76925-830 76925-832
5-Ethyl-2-fluorophenylboronic acid ≥95%
5-Ethyl-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD18783131
CAS Number: 900175-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR