4-Bromo-l-chloro-2-ethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

16540-25G 16540-5G
76925-820EA 418 USD
76925-820 76925-822
4-Bromo-l-chloro-2-ethoxybenzene ≥97%
4-Bromo-l-chloro-2-ethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₈BrClO
Storage Temperature: Refrigerator
MDL Number: MFCD12547812
CAS Number: 900174-61-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR