4-Carbamoyl-2-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Carbamoyl-2-fluorophenyl)boronic acid

16531-1G 16531-25G 16531-500MG 16531-5G
76925-808EA 135.8 USD
76925-808 76925-810 76925-812 76925-814
4-Carbamoyl-2-fluorophenylboronic acid ≥97%
4-Carbamoyl-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BFNO₃
MW: 182.9500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09800867
CAS Number: 874289-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR