4-Methoxy-1-naphthaleneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Methoxynaphthalen-1-yl)boronic acid, 4-Methoxynaphthalene-1-boronic acid

16377-25G 16377-5G
76925-708EA 434.97 USD
76925-708 76925-710
4-Methoxy-1-naphthaleneboronic acid ≥97%
4-Methoxy-1-naphthaleneboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD03452757
CAS Number: 219834-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR