5-Bromo-2,4-dimethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16234-25G 16234-5G
76925-698EA 230.22 USD
76925-698 76925-700
5-Bromo-2,4-dimethoxybenzaldehyde ≥97%
5-Bromo-2,4-dimethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
Storage Temperature: Ambient
MDL Number: MFCD00010863
CAS Number: 130333-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR